Sodium phosphate, monobasic dihydrate, 98%, extra pure
Sodium phosphate, monobasic dihydrate, 98%, extra pure
Sodium phosphate, monobasic dihydrate, 98%, extra pure
Acros Organics

Sodium phosphate, monobasic dihydrate, 98%, extra pure

CAS: 13472-35-0 | H2NaO42H2O | 156.01 g/mol
Have Questions?
Catalog NumberQuantity
447760050
also known as 44776
5 kg
Catalog number 447760050
also known as 44776
HSN 28353900
Price (INR)
22,512.00
Each
Quantity:
5 kg
Request bulk or custom format
Price (INR)
22,512.00
Each
Chemical Identifiers
CAS13472-35-0
IUPAC Namesodium dihydrate dihydrogen phosphate
Molecular FormulaH6NaO6P
InChI KeyVBJGJHBYWREJQD-UHFFFAOYSA-M
SMILESO.O.[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Crystals
Titration with NaOH97.5 to 102.5 %
pH4.2 to 4.5 (5%, water)
Loss on drying21.5 % to 24.0 % (130°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only