Sodium sulfite, 98%, for biochemistry
Sodium sulfite, 98%, for biochemistry
Sodium sulfite, 98%, for biochemistry
Acros Organics

Sodium sulfite, 98%, for biochemistry

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol
Have Questions?
Catalog NumberQuantity
447950010
also known as 44795
1 kg
Catalog number 447950010
also known as 44795
HSN 28321010
Price (INR)
19,931.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
19,931.00
Each
Chemical Identifiers
CAS7757-83-7
IUPAC Namedisodium heptahydrate sulfite
Molecular FormulaH14Na2O10S
InChI KeyWMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Iodimetric>=97.5 %
Solubility(1 M in water) Clear colorless
Chloride (Cl)=<0.02 %
Copper (Cu)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only