QuantStudio™ Dx Real-Time PCR Instrument with array card block
Applied Biosystems™

QuantStudio™ Dx Real-Time PCR Instrument with array card block

The QuantStudio™ Dx Real-Time PCR Instrument with Array Card Block is a real-time PCR platform that delivers the proven performance,Read more
Have Questions?
Catalog NumberQuantity
44798911 Instrument
Catalog number 4479891
Price (INR)
-
Quantity:
1 Instrument
The QuantStudio™ Dx Real-Time PCR Instrument with Array Card Block is a real-time PCR platform that delivers the proven performance, flexibility, expandability, and support needed to meet the evolving needs of the clinical laboratory today and tomorrow. The QuantStudio™ Dx Real-Time PCR Instrument with Array Card Block offers easy-to-change blocks that can also accommodate 96-well fast, 96-well standard, and 384-well plates, allowing you to select the format that is right for your throughput needs.

Built on a legacy of innovation and excellence in real-time PCR, the QuantStudio™ Dx instrument leverages the best of our portfolio of Applied Biosystems™ real-time PCR systems, and refines a platform to specifically meet the needs of the diagnostic laboratory.

Results You Can Trust
The QuantStudio™ Dx Real-Time PCR Instrument’s proven performance, security, and reliability—combined with the outstanding sensitivity and specificity of the Quidel™ Molecular Assays—help ensure the right answer is delivered the first time, every time.

Flexibility You Want, Security You Need
The QuantStudio™ Dx Real-Time PCR Instrument with Array Card Block is designed with easily interchangeable thermal cycling blocks to accommodate your laboratory’s varying needs for throughput, format type, and assay type. In addition to the array card format, the instrument also accepts 96-well fast, 96-well standard and 384-well card formats that can be used for laboratory-developed tests (LDTs) and future IVD assays. Additionally, the system can be operated in Test Development Mode, which enables clinical laboratories to develop their own assays or perform clinical research.

Efficient Workflows and Assays
A simplified instrument and assay workflow as well as intuitive software make the entire test easy and efficient, from processing to reporting. Plus, it frees up highly trained operators to perform more complex tasks.

QuantStudio™ Dx Software
• Track reagent and patient information for quality control purposes
• Enter reagent information by barcode and upload patient information from Excel files to minimize data entry errors
• Print calibration and maintenance reports for your instrument records
• Included SAE module assists with Security, Auditing, and Electronic signature of records for full data traceability as part of 21 CRF Part 11 compliance

For In Vitro Diagnostic Use. The QuantStudio™ Dx Real-Time PCR Instrument meets the requirements of the in vitro Diagnostic Medical Devices Directive (98/79/EC). The CE IVD-registered QuantStudio™ Dx Real-Time PCR Instrument is for distribution and use in specific countries only. Not available for sale in the United States.
Specifications
Calibrated DyeFAM™, VIC™, ROX™, TAMRA, CY™5
Detection MethodFluorescence
Dimensions53.0 cm (20.9 in.) W x 70.0 cm (27.6 in.) D x 75.0 cm (29.5 in.) H
Dynamic RangeUp to 9 Logs of Linear Dynamic Range
For Use With (Equipment)QuantStudio™ Dx
Format384-well Microfluidic Card
Optics6 Emission Filters (500-720 nm), 6 Excitation Filters (450-670 nm)
Product LineQuantStudio
Quantity1 Instrument
Reaction SpeedStandard
Reaction Volume Range1 μl
SensitivityDown to 1 Copy
Software CompatibilityQuantStudio™ Dx Software; QuantStudio™ Test Development Software
Thermal Uniformity±0.5°C
Weight70 kg
Block ConfigurationsArray Card
Block FormatInterchangeable
Capacity1 Array Card
Unit SizeEach
Contents & Storage
This instrument comes with an Array Card Block.

Frequently asked questions (FAQs)

What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

I recently switched the block in my real-time PCR instrument. Now it shows the status as 'not calibrated' for all the calibration plates, even though my calibrations are current. What is causing this issue?

The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:

  1. Remove the block from the instrument.
  2. Reinsert the block into the instrument.


If the problem persists after reseating the block, please contact our Technical Support team for further assistance.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.