MicroAmp™ GPLE Adhesive Film Applicators
Applied Biosystems™
MicroAmp™ GPLE Adhesive Film Applicators
Applied Biosystems™ MicroAmp™ GPLE Adhesive Film Applicators form a tight seal between a microplate and an adhesive film. Each lot of these General Purpose Laboratory Equipment (GPLE) applicators is manufactured in an ISO-registered facility.
Have Questions?
Catalog NumberQuantity
44811965 Applicators
Catalog number 4481196
Price (INR)
11,466.00
Online offer
Ends: 31-Dec-2026
19,110.00
Save 7,644.00 (40%)
Each
Quantity:
5 Applicators
Price (INR)
11,466.00
Online offer
Ends: 31-Dec-2026
19,110.00
Save 7,644.00 (40%)
Each
Use an Applied Biosystems MicroAmp GPLE Adhesive Film Applicator to form a tight seal between a microplate and an adhesive film. Each lot of these General Purpose Laboratory Equipment (GPLE) applicators is manufactured in an ISO-registered facility.
Form a tight seal
The MicroAmp GPLE Adhesive Film Applicator enables you to ergonomically apply pressure to an adhesive film upon a microplate to form a tight seal and minimize air bubbles. This tight seal can help prevent well-to-well contamination and sample loss.
Quick and easy
Use the MicroAmp GPLE Adhesive Film Applicator to speed the process of sealing multiple microplates. This tool is designed according to the microplate dimensions so you can quickly seal a microplate. The rugged proprietary plastic endures constant use so you can complete large screening applications.
Note: See user's manual or package insert for limited label license and trademark information.
For Laboratory Use
Specifications
ColorGray
MaterialPolyethylene
No. per Pack5 Applicators
Quantity5 Applicators
Product LineMicroAmp
Product TypeFilm, Foil, or Seal Applicator
Unit SizeEach