Product Image
Applied Biosystems™

Background Plate, 96-well

The 96-Well Background Plate is used to maintain your real-time PCR instrument with 96-well standard block. Background calibration should beRead more
Have Questions?
Catalog NumberQuantity
44813091 x 96 well plate
Catalog number 4481309
Price (INR)
15,313.00
Each
Quantity:
1 x 96 well plate
Price (INR)
15,313.00
Each
The 96-Well Background Plate is used to maintain your real-time PCR instrument with 96-well standard block. Background calibration should be run at least every six months. Please see your instrument user guide for full instructions on use.
Specifications
For Use With (Equipment)ViiA™ 7 Dx System, QuantStudio™ Dx
Packaging1 x 96 Plate
Product TypeBackground plate
Quantity1 x 96 well plate
No. of Wells96
Unit SizeEach
Contents & Storage
Store at –20°C ± 5°C.