OpenArray® 384-well Sample Plates are for use with the QuantStudio™ 12K Flex Real-Time PCR System. They are low-volume microplates usedRead more
Have Questions?
Catalog Number
Quantity
4482221
10 plates
Catalog number 4482221
Price (INR)
19,591.00
Each
Quantity:
10 plates
Price (INR)
19,591.00
Each
OpenArray® 384-well Sample Plates are for use with the QuantStudio™ 12K Flex Real-Time PCR System. They are low-volume microplates used in the QuantStudio™ OpenArray® Accufill™ System for sample loading onto OpenArray® plates.
These particular plates are black in color for improved visualization during loading.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BarcodeNo Barcode
ColorBlack
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)QuantStudio™
Format384-well Plate
Product LineOpenArray
Quantity10 plates
Reaction SpeedStandard
Product TypeSample Plate
Unit SizeEach
Contents & Storage
Each box contains 10 x 384-well plates, enough for 80 OpenArray™ Real-Time PCR plates.