DS-33 GeneScan™ Install Kit Dx with GeneScan™ 600 LIZ™ Size Standard v2.0 Dx
Product Image
Applied Biosystems™
DS-33 GeneScan™ Install Kit Dx with GeneScan™ 600 LIZ™ Size Standard v2.0 Dx
Developed for use with Applied Biosystems fluorescence-based DNA electrophoresis systems. The use of internal size standard enables automated data analysis and is also essential for achieving high run to run precision in sizing DNA fragments.
Have Questions?
Catalog NumberQuantity
44829751 Kit
Catalog number 4482975
Price (INR)
-
Quantity:
1 Kit
The DS-33 GeneScan Install Kit Dx w/ GeneScan 600 LIZ Size Standard v2.0 Dx consists of pooled PCR products labeled with 6-FAM, VIC, NED, and PET dyes. To generate the pooled products, CEPH individual 1347-02 genomic DNA is amplified with eight fluorescent PCR primer pairs, which amplify selected microsatellite loci (as listed in the package product insert). The kit also contains a tube of GeneScan 600 LIZ Size Standard v2.0 Dx (Cat. No. A25794).
For In Vitro Diagnostic Use Only
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3500 Dx Genetic Analyzer
Label or DyeLIZ
Product LineGeneScan, LIZ
Product TypeInstall Kit
Quantity1 Kit
Shipping ConditionDry Ice
FormatKit
Unit SizeEach
Contents & Storage
Each kit contains four tubes of GeneScan Installation Standard DS-33 and one tube of GeneScan 600 LIZ size standard v2.0 Dx.
Store the kit at -15 to -25°C until ready to use. Upon thawing, store the GeneScan 600 LIZ Size Standard v2.0 tube at 2–8°C.