MicroAmp™ EnduraPlate™ Optical 384-Well Clear GPLE Reaction Plates with Barcode
Applied Biosystems™ MicroAmp™ EnduraPlate™ GPLE plastic consumables provide superior PCR and qPCR performance and quality in formats developed to meetRead more
Have Questions?
Catalog Number
Quantity
4483319
20 plates
Catalog number 4483319
Price (INR)
13,938.00
Online offer
Ends: 31-Dec-2026
23,229.00
Save 9,291.00 (40%)
Each
Quantity:
20 plates
Price (INR)
13,938.00
Online offer
Ends: 31-Dec-2026
23,229.00
Save 9,291.00 (40%)
Each
Applied Biosystems™ MicroAmp™ EnduraPlate™ GPLE plastic consumables provide superior PCR and qPCR performance and quality in formats developed to meet your experimental needs. All EnduraPlate™ GPLE plastic consumables are validated on Applied Biosystems™ thermal cyclers, real-time thermal cyclers, and genetic analyzers for optimal fit and performance.
Key Features and Benefits:
• Each lot is certified DNA, RNAse, and PCR inhibitor-free • Labeled 'For Laboratory Use' • Optimized for performance with Applied Biosystems™ instruments • Excellent mechanical stability and flatness for your complete workflow needs
Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate offers mechanical stability, flatness, and excellent durability. ANSI/SBS-compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate™ reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler → real-time thermal cycler → genetic analyzer).
EnduraPlate™ reaction plates have a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample. These plates are optimized for performance with Applied Biosystems™ instruments. Easy-to-read, black, well-identification text helps to minimize pipetting mistakes and improves sample preparation productivity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Packaging20 Plates
Product TypeMicroplate
Quantity20 plates
Reaction SpeedStandard
Skirt StyleSkirted
BarcodeIncludes Barcode
ColorOptical, Clear
MaterialPolycarbonate, Polypropylene
No. of Wells384
Product LineMicroAmp
Unit SizeEach
Contents & Storage
20 384-Well Fast plates. Store at room temperature