MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates with Barcode
Applied Biosystems™ MicroAmp™ EnduraPlate™ plastic consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs.Read more
Have Questions?
Change view
Catalog Number
Quantity
4483352
500 Plates
4483354
20 Plates
2 Options
Catalog number 4483352
Price (INR)
528,623.00
Each
Quantity:
500 Plates
Price (INR)
528,623.00
Each
Applied Biosystems™ MicroAmp™ EnduraPlate™ plastic consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. All of our plastic consumables are validated with Applied Biosystems™ instruments for optimal fit and performance. With the new EnduraPlate™ reaction plates, there is now a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments.
Key Features and Benefits:
• Optimized for performance with Applied Biosystems™ instruments • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests • Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling • Available in multiple colors for easy organization of laboratory assays • Excellent mechanical stability and flatness for your complete workflow needs
Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability. ANSI/SBS compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate™ reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler -> real-time thermal cycler -> genetic analyzer).
EnduraPlate™ reaction plates have a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample. These plates are optimized for performance with Applied Biosystems™ instruments. Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity, while five color choices provide a convenient way to organize your laboratory assays.
Note: An adapter (Cat No. A24820) is required for use of this product with 7300/7500 Real-Time PCR systems.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Packaging500 Plates
Product TypeMicroplate
Quantity500 Plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Well0.3 mL
Volume (Metric) Working0.2 mL
BarcodeIncludes Barcode
ColorClear, Optical
MaterialPolycarbonate, Polypropylene
No. of Wells96
Product LineMicroAmp
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
What is included in Oncomine Precision Assay GX (Cat. No. A46291)?
Oncomine Precision Assay GX (Cat. No. A46291) includes the following:
- OPA Pool 1 panel (Part No. A44350)
- Genexus Strip 1, quantity of 8 strips (Part No. A46812)
- Genexus Strip 2-HD, quantity of 8 strips (Part No. A46814)
- The OPA Pool 1 is provided in 8 pairs of tubes, where each tube pair contains one tube with forward (FWD) primers and one tube with reverse (REV) primers.