TaqMan™ Verotoxin-producing Escherichia coli VT1/VT2 Multiplex Assay, HPA
Thermo Scientific™

TaqMan™ Verotoxin-producing Escherichia coli VT1/VT2 Multiplex Assay, HPA

This TaqMan™ assay provides a simple, reliable, and rapid method for detecting the presence of a specific bacterial or viralRead more
Have Questions?
Catalog NumberQuantity
448507496 reactions
Catalog number 4485074
Price (INR)
-
Quantity:
96 reactions
This TaqMan™ assay provides a simple, reliable, and rapid method for detecting the presence of a specific bacterial or viral target. The assay uses PCR to amplify a DNA target unique to the microorganism and TaqMan™ probe to detect and quantify the target, all in an an easy-to-use, lyophilized format. TaqMan™ assays deliver the fastest RT-PCR run times on the market (<40 minutes in Fast mode). Simply add 30 μL of purified DNA to the assay tube and run on Fast or standard instrumentation. The TaqMan™ assay provides microbial detection capabilities and high-confidence results to help accelerate your food and environmental testing needs. The assay design is based on a publication by Nielsen, et al. [Nielsen EM, Andersen MT (2003) J Clin Micro 41:2884-2983] and validated by the Health Protection Assocation (HPA).

Features of the TaqMan™ assay:

• Convenience—simply add your nucleic acid sample to the assay tube and run; no pre-mixing or master mix preparation required
• Consistency—fewer operator steps means reduced error and increased reproducibility across time, users, and laboratories
• Improved data accuracy—an internal positive control is included to monitor your PCR
• Fast chemistry—40-min run time with Fast cycling, compatible with standard or Fast modes

TaqMan™ assays are suitable for single or multiplex analysis. Because they are run using universal conditions, multiple assays can be analyzed in the same run. The robust assay design provides highly accurate, reproducible, and reliable results, with the enhanced stability of a lyophilized format. A passive internal reference based on proprietary ROX™ dye is included for precise data analysis.

This product is for environmental testing, quality control/quality assurance testing, food and agricultural testing, and research purposes only.
Specifications
DescriptionIncludes Master Mix
No. of Reactions96 Reactions
Quantity96 reactions
SpeciesAll Species
Target Organism ClassE. coli
Detection MethodPrimer-probe
For Use With (Application)Food and Beverage
FormLiquid
Reaction SpeedStandard
Unit Size96 reactions
Contents & Storage
• 12 8-tube strips (store at 2–8°C in the dark)
• Optical 8-tube strips and 8-cap strips (store at room temperature)

These reagents are sufficient for 96 reactions.

Citations & References (1)

Citations & References
Abstract
Evaluation of seven different commercially available real-time PCR assays for detection of shiga toxin 1 and 2 gene subtypes.
Authors:Margot H, Cernela N, Iversen C, Zweifel C, Stephan R,
Journal:J Food Prot
PubMed ID:23643131
Following the recent outbreak of Shiga toxin-producing Escherichia coli (STEC) O104:H4 infection in Germany, the demand for fast detection of STEC has again increased. Various real-time PCR-based methods enabling detection of Shiga toxin genes (stx) have been developed and can be used for applications in food microbiology. The present study ... More