NUCLEIC-CARD™ Collection Devices, framed
Thermo Scientific™
NUCLEIC-CARD™ Collection Devices, framed
NUCLEIC-CARD is an easy-to-use system for collection, preservation, and long-term storage of nucleic acids at room temperature. The NUCLEIC-CARD matrixRead more
Have Questions?
Catalog NumberQuantity
4486610CBox of 50
Catalog number 4486610C
Price (INR)
-
Quantity:
Box of 50
NUCLEIC-CARD is an easy-to-use system for collection, preservation, and long-term storage of nucleic acids at room temperature. The NUCLEIC-CARD matrix is chemically treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature.
Features of NUCLEIC-CARD include:
  • Enables direct PCR amplification from a card punch, eliminating extraction steps
  • Facilitates high quality STR profiles with Applied Biosystems direct PCR amplification kits
  • Certified free of DNase, RNase, and amplifiable human DNA
  • Prevents growth of microorganisms, enabling long-term room-temperature storage
  • Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity
Traceability
Every NUCLEIC-CARD is identified with a pre-printed barcode.
Automation and cleaning area
NUCLEIC-CARD is compatible with CPA200 and other punching systems and includes an area for cleaning strikes while punching with manual or automated punchers.
NUCLEIC-CARD is manufactured by Copan, a market leader in the area of sample collection tools, and distributed by Thermo Fisher Scientific.
NUCLEIC-CARD is a trademark of Copan Flock Technologies.
For Criminal Justice and Forensic Use Only.
Specifications
Dimensions (L x W)25.4 x 25.4 mm (Depot area), 51 x 51 mm (Total)
For Use With (Application)Collection, preservation, and long-term storage of nucleic acids at room temperature
QuantityBox of 50
Product TypeNUCLEIC-CARD™
Unit SizeEach