Doxylamine succinate, 98%
Doxylamine succinate, 98%
Doxylamine succinate, 98%
Doxylamine succinate, 98%
Acros Organics

Doxylamine succinate, 98%

CAS: 562-10-7 | C21H28N2O5 | 388.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
449540500
also known as 44954
50 gGlass bottle
Catalog number 449540500
also known as 44954
HSN 29333919
Price (INR)
140,869.00
Each
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
140,869.00
Each
Add to cart
Chemical Identifiers
CAS562-10-7
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point103°C to 108°C
Appearance (Color)White to off-white
Titration with HClO4>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only