Product Image
Thermo Scientific™

TaqMan™ TAMRA™ PROBE, 5'FAM™

Custom TaqMan™ TAMRA™ Probes are dual labeled, real-time PCR probes that can be used in all of your real-time PCRRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4500030150 nmol
4500250110 nmol
4500240120 nmol
Catalog number 45000301
Price (INR)
-
Quantity:
50 nmol
Custom TaqMan™ TAMRA™ Probes are dual labeled, real-time PCR probes that can be used in all of your real-time PCR applications.

• Choice of synthesis scales
• Choice of powder or liquid format
• Available for any species or organism
• For use in quantitative gene expression, pathogen detection, and other real time PCR applications

Applied Biosystems Custom TaqMan™ TAMRA™ Probes incorporate a 5’ reporter dye and a 3’ TAMRA™ quencher dye. All Custom TaqMan™ TAMRA™ Probes are HPLC purified. Design Custom TaqMan™ TAMRA™ Probes for any species, splice variant or novel gene. Custom TaqMan™ TAMRA™ probes are available with the following reporter dyes; FAM™, VIC™.

Convenient Ordering and Delivery
Now some Custom TaqMan™ MGB™ Probes are manufactured in China, with exactly the same quality as those made in US, while the turnaround time shorten to 5-7 working days. TaqMan™ MGB Probes have a shelf life of 12 months from manufacturing date.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Contact Us
Please download the electronic order file, and send it to synsh@invitrogen.com.

Hotline: 800-820-8982 / 400-820-8982
Specifications
Product LineTaqMan
Product TypeTAMRA Probe
Purity or Quality GradeHPLC
Quantity50 nmol
Shipping ConditionApproved for shipment on Wet or Dry Ice
For Use With (Application)Real Time PCR (qPCR)
FormPowder, Liquid
Unit SizeEach
Contents & Storage
-20°C

Frequently asked questions (FAQs)

How can I calculate the concentration of a TaqMan probe?

The initial concentration or total yield of your synthesized probe is listed on the data sheet supplied with the probe. TaqMan probes are sometimes shipped in the lyophilized state, but more often are shipped in solution (1X TE). To create a working stock solution of your probe or primers, you should reconstitute or dilute the probe in the appropriate volume of sterile 1X TE (1 mM Tris, 0.1 mM EDTA, pH 8.0) or sterile, nuclease-free H2O. Detailed instructions on how to calculate the proper volume of solution to add to your probe are provided in our online Tutorial document "Reconstituting and Diluting Primers and TaqMan Probes". You can find a copy on our website by entering this title in the main Search field.

Why does a TaqMan TAMRA probe need to have more C's (Cytosines) than G's (Guanidines) in the sequence?

Empirical data has shown that TaqMan TAMRA probes which contain more Gs than Cs will generally have lower ΔRn values and overall poor performance compared to those that do not. For more information on designing TaqMan TAMRA probes, please consult tutorials related to the Primer Express software.

What are your recommendations for the best way to dilute and store my TaqMan probe?

The probe should be diluted in TE buffer [10 mM Tris-HCl, pH 8.0 (at 25°C), 1 mM EDTA]. We recommend storing your probes, both stocks and working solutions, at -20°C in aliquots. This is to cut down on the number of times the probe is subject to freeze-thaws. For detailed information on diluting primers and probes, please refer to the tutorial entitled "Reconstituting and Diluting Primers and TaqMan Probes". You can find it on our website by entering this title in the main search field.

Find additional tips, troubleshooting help, and resources within ourTaqMan Primers and Probes Support Center.