Product Image
Thermo Scientific™

TaqMan™ TAMRA™ PROBE, 5'VIC™, 20 nmoles

Custom TaqMan™ TAMRA™ Probes are dual labeled, real-time PCR probes that can be used in all of your real-time PCRRead more
Have Questions?
Catalog NumberQuantity
450024021 tube
Catalog number 45002402
Price (INR)
-
Quantity:
1 tube
Custom TaqMan™ TAMRA™ Probes are dual labeled, real-time PCR probes that can be used in all of your real-time PCR applications.

• Choice of synthesis scales
• Choice of powder or liquid format
• Available for any species or organism
• For use in quantitative gene expression, pathogen detection, and other real time PCR applications

Applied Biosystems Custom TaqMan™ TAMRA™ Probes incorporate a 5’ reporter dye and a 3’ TAMRA™ quencher dye. All Custom TaqMan™ TAMRA™ Probes are HPLC purified. Design Custom TaqMan™ TAMRA™ Probes for any species, splice variant or novel gene. Custom TaqMan™ TAMRA™ probes are available with the following reporter dyes; FAM™, VIC™.

Convenient Ordering and Delivery

Now some Custom TaqMan™ MGB™ Probes are manufactured in China, with exactly the same quality as those made in US, while the turnaround time shorten to 5-7 working days. TaqMan™ MGB Probes have a shelf life of 12 months from manufacturing date.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Contact Us

Please download the electronic order file, and send it to synsh@invitrogen.com.

Hotline: 800-820-8982 ⁄ 400-820-8982
Specifications
FormatTube
Product LineTaqMan
Product TypeTAMRA Probe
Purity or Quality GradeHPLC
Quantity1 tube
Shipping ConditionApproved for shipment on Wet or Dry Ice
For Use With (Application)Real Time PCR (qPCR)
FormPowder, Liquid
Unit SizeEach
Contents & Storage
-20°C