Injection Port Liners for GC Instruments
Actual product may vary
Injection Port Liners for GC Instruments
Use these standard deactivated injection port liners to optimize performance of Thermo Scientific and Agilent GC instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerPacking MaterialProduct TypeQuantity
453A12954 mmNoneSplit Straight Liner5
453A12524 mmQuartz WoolSplit/Splitless FocusLiner25
453A12554 mmQuartz WoolSplit/Splitless FocusLiner5
453A12624 mmQuartz WoolSplit Straight Liner25
453A12852.3 mmQuartz WoolSplit/Splitless FAST FocusLiner5
453A13054 mmQuartz WoolSplit/Splitless Liner w/ Recessed Gooseneck5
453A13124 mmQuartz WoolSplit/Splitless FocusLiner w/ Single Taper25
453A13154 mmQuartz WoolSplit/Splitless FocusLiner w/ Single Taper5
453A13351.2 mmNoneDirect Straight Liner5
453A13454 mmNoneSplit/Splitless Liner w/ Single Taper5
453A13554 mmNoneSplit/Splitless Liner w/ Double Taper5
453A19254 mmQuartz WoolSplitless Liner w/ Single Taper5
453A20015.2 mmNoneSingle Gooseneck Liner5
453A20025.2 mmNoneCyclosplitter Liner5
453A20035.2 mmQuartz WoolSplit/Splitless Liner5
453A22654 mmQuartz WoolSplit Straight Liner5
453A22752 mmQuartz WoolSplitless Straight Liner5
453A22822.3 mmQuartz WoolSplit/Splitless FAST FocusLiner25
453A22924 mmSplit Straight Liner for Agilent Instruments25
453A23424 mmNoneSplit/Splitless Liner w/ Single Taper25
453A23524 mmNoneSplit/Splitless Liner w/ Double Taper25
453A23752.3 mmNoneSplit/Splitless FAST FocusLiner w/ Single Taper5
453A2295
also known as 453A2295 TRAJAN SCIENTIFIC AMERICAS INC
4 mmQuartz WoolSplitless Liner w/ Single Taper25
Catalog number 453A1295
HSN 84212900
Price (INR)
14,468.00
Pack
Diameter (Metric) Inner:
4 mm
Packing Material:
None
Product Type:
Split Straight Liner
Quantity:
5
Price (INR)
14,468.00
Pack
Optimize the performance of Thermo Scientific™ and Agilent™ GC systems with our specifically designed Thermo Scientific™ Injection Port Liners for Agilent instruments. Highly deactivated and produced to exacting tolerances, these injection liners ensure exceptional reproducibility.
  • Highly deactivated borosilicate glass
  • Quartz wool packing material
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Split
Quantity5
Length (Metric)78.5 mm
Packing MaterialNone
Product TypeSplit Straight Liner
Diameter (Metric) Inner4 mm
Diameter (Metric) Outer6.3 mm
For Use With (Equipment)Thermo Scientific TRACE 1300/1600 series and Agilent GC instruments
Liner MaterialHighly Deactivated Borosilicate Glass
Unit SizePack