Injection Port Liners with CarboFrit for Thermo Scientific™ Trace and Focus SSL Instruments
Actual product may vary
Injection Port Liners with CarboFrit for Thermo Scientific™ Trace and Focus SSL Instruments
Ensure a high degree of reproducibility with Thermo Scientific Injection Port Liners with CarboFrit for Thermo Scientific Trace and Focus SSL Instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
453T2131Split Straight Liner
453T2130Splitless Straight Liner
Catalog number 453T2131
HSN 84212900
Price (INR)
73,891.00
Pack
For Use With (Equipment):
Split Straight Liner
Price (INR)
73,891.00
Pack
Ensure a high degree of reproducibility with Thermo Scientific Injection Port Liners with CarboFrit for Thermo Scientific Trace and Focus SSL Instruments. Highly deactivated and produced to exacting tolerances.
Specifications
Description453T2131
Quantity5
Length (Metric)105 mm
Packing MaterialCarboFrit
Product TypeSplit Straight Liner
Diameter (Metric) Inner5 mm
Diameter (Metric) Outer8 mm
For Use With (Equipment)Split Straight Liner
Unit SizePack