TCF-32 Continuous-Flow Rotors
Thermo Scientific™
TCF-32 Continuous-Flow Rotors
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of these rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityDescription
45835430mL430mL Standard Core
45820940mL/430 mLTCF-32 Continuous-Flow Rotor
45830940 mL940mL Accessory Core
Catalog number 45835
Price (INR)
-
Capacity:
430mL
Description:
430mL Standard Core
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of Thermo Scientific™ TCF-32 Continuous-Flow Rotors.
Compatible with:
Thermo Scientific Sorvall WX Ultra Centrifuge Series
Ordering Information:
Core is required for operation. Must be purchased separately.
Specifications
Description430mL Standard Core
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
K Factor42
Capacity430mL
Max. RCF102,000 x g
Max. Speed32,000 rpm
TypeContinuous Flow Rotor
Unit SizeEach