DyLight™ 405 Maleimide
Thermo Scientific™

DyLight™ 405 Maleimide

Thermo Scientific DyLight 405 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 405 used to fluorescently label sulfhydryl-containing peptides,Read more
Have Questions?
Catalog NumberQuantity
466001 mg
Catalog number 46600
Price (INR)
-
Quantity:
1 mg
Thermo Scientific DyLight 405 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 405 used to fluorescently label sulfhydryl-containing peptides, proteins and other biomolecular probes.

DyLight 405 has high fluorescence intensity over a broad pH range (pH 4-9) and is more photostable than Alexa Fluor™ 405, Cascade™ Blue and AMCA dyes in many applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates.

Features of DyLight 405 Maleimide:

High performance—DyLight 405 shows brighter fluorescence than Alexa Fluor 405, Cascade Blue and AMCA
Specific—maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
Efficient labeling methods—well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
Optimized antibody labeling procedure—complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
• Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
• Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies

DyLight 405 Sulfhydryl-Reactive Dye is activated with a maleic acid imide (maleimide) moiety to form a reactive alkylation reagent. Labeling occurs through reaction of the maleimide-activated dye with reduced sulfhydryl groups (-SH) to form stable thioether bonds. Maleimides are specific for sulfhydryl groups between pH 6.5-7.5. Learn more about maleimide chemistry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
DescriptionDyLight 405 Maleimide
Emission420
Excitation400
Label or DyeDyLight™ 405
Product TypeDye
Quantity1 mg
Reactive MoietyMaleimide
SolubilityDMF (Dimethylformamide)
Label TypeDyLight Fluors
Product LineDyLight
Unit SizeEach
Contents & Storage
Store desiccated at -20°C. Shipped at ambient temperature.