DyLight™ 800 Maleimide
Thermo Scientific™

DyLight™ 800 Maleimide

Thermo Scientific DyLight 800 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 800 used to fluorescently label sulfhydryl-containing peptides,Read more
Have Questions?
Catalog NumberQuantity
466211 mg
Catalog number 46621
Price (INR)
30,900.00
線上優惠
Ends: 31-Dec-2026
51,500.00
Save 20,600.00 (40%)
Each
Quantity:
1 mg
Price (INR)
30,900.00
線上優惠
Ends: 31-Dec-2026
51,500.00
Save 20,600.00 (40%)
Each
Thermo Scientific DyLight 800 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 800 used to fluorescently label sulfhydryl-containing peptides, proteins and other biomolecular probes.

DyLight 800 is a near-IR fluor that is invisible to the naked eye but increases the staining options when using infrared imaging systems. DyLight 800 has spectral properties that are very similar to other near-IR dyes, including Alexa Fluor™ 790 and IRDye™ 800. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates.

Features of DyLight 800 Maleimide:

• High performance—DyLight 800 replaces Alexa Fluor 800 and IRDye 800 for near-infrared staining
• Specific—maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
• Efficient labeling methods—well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
• Optimized antibody labeling procedure—complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
• Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting, and ELISA assay
• Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies

DyLight 800 Sulfhydryl-Reactive Dye is activated with a maleic acid imide (maleimide) moiety to form a reactive alkylation reagent. Labeling occurs through reaction of the maleimide-activated dye with reduced sulfhydryl groups (-SH) to form stable thioether bonds. Maleimides are specific for sulfhydryl groups between pH 6.5-7.5. Learn more about maleimide chemistry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
DescriptionDyLight 800 Maleimide
Emission794
Excitation777
Label or DyeDyLight™ 800
Product TypeDye
Quantity1 mg
Reactive MoietyMaleimide
SolubilityDMF (Dimethylformamide)
Label TypeDyLight Fluors
Product LineDyLight
Unit SizeEach
Contents & Storage
Store desiccated at -20°C.