Barcoded Latch Racks for Matrix™ Storage Tubes
Actual product may vary

Barcoded Latch Racks for Matrix™ Storage Tubes

Create custom arrays with empty ANSI-format racks featuring 2D and linear barcodes and human readable type on three sides and a 2D barcode on the base of the rack.
Have Questions?
Catalog NumberFor Use With (Equipment)No. per Case
4900-BR500μL Screwtop Tubes5
Catalog number 4900-BR
Price (INR)
15,160.00
Each
For Use With (Equipment):
500μL Screwtop Tubes
No. per Case:
5
Price (INR)
15,160.00
Each
Enhance sample storage at low temperatures, including vapor phase Liquid Nitrogen, with Thermo Scientific™ Matrix™ barcoded latch racks. Barcoded Latch Racks feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack for the ultimate in sample tracking. Use in combination with Thermo Scientific 2D barcoded tubes, barcode readers, arrayers and capping equipment.

Superior Storage Rack Design

  • Storage in robot-friendly ANSI-footprint rack
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking regardless of your barcode system
  • Rack barcodes are compatible with our range of VisionMate readers
  • Hinged lid rests in open position on rack while conducting manual pipetting to prevent contamination of lid sitting on bench

Latch Racks Save Precious Space

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Latch rack lid can be lifted robotically for tube access with high-throughput automated applications
Specifications
ColorWhite
DescriptionEmpty Latch Rack
Capacity (Metric)0.5 mL
MaterialPlastic
For Use With (Equipment)500μL Screwtop Tubes
No. per Case5
Barcode2D Datamatrix; Linear EAN Code 128; Human Readable
Holds2D Matrix tubes
Product LineMatrix
Packaging Quantity5 Case
Capacity (English)0.0169 fl oz.
Unit SizeEach