Four Inner-Door Option for Thermo Scientific™ Ultra-Low Temperature Freezers
Thermo Scientific™
Four Inner-Door Option for Thermo Scientific™ Ultra-Low Temperature Freezers
Use with Thermo Scientific™ Revco™ UxF and HERAfreeze™ HFU T Series freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
4D300HERAfreeze HFU300T, Revco UxF300
4D400HERAfreeze HFU400T, Revco UxF400
4D500HERAfreeze HFU500T, Revco UxF500
4D600HERAfreeze HFU600T, Revco UxF600
4D700HERAfreeze HFU700T, Revco UxF700
Catalog number 4D300
Price (INR)
-
For Use With (Equipment):
HERAfreeze HFU300T, Revco UxF300
Choose one of the Thermo Scientific™ Four Inner-Door Options Ultra-Low Temperature Freezers for your lab. Use these doors with Thermo Scientific™ Revco™ UxF and HERAfreeze™ HFU T Series freezers.
Specifications
For Use With (Equipment)HERAfreeze HFU300T, Revco UxF300
TypeUltra-Low Freezer Inner-Door
No. of Doors4
Unit SizeEach