Adjust speed from 100 to 1000rpm with this external control unit required for use with the Mobil 10 and Mobil 25 stirrers.
Have Questions?
Catalog Number
Voltage
50119121
115 V
Catalog number 50119121
Price (INR)
-
Voltage:
115 V
Adjust speed from 100 to 1000rpm with the Thermo Scientific™ Cimarec™ 10M Controller, an external control unit that is required for use with the Mobil 10 and Mobil 25 stirrers.
Automatic start-up for safe stirrer acceleration
Specifications
Electrical Requirements115 V
For Use With (Equipment)Mobil 10 and Mobil 25 Stirrer