Product Image
Thermo Scientific™

Cytomat™ 10 Hotel Ambient Storage (Perspex Housing), Stainless Steel

Temperature Range: Ambient
Have Questions?
Catalog NumberMaterial
50139108Stainless Steel
Catalog number 50139108
Price (INR)
-
Material:
Stainless Steel

Designed to fit into the busiest of cell biology or screening laboratories, the Thermo Scientific™ Cytomat™ 10 Hotel ambient storage is the ideal solution for high‐capacity random access storage (210 SBS microplates) at ambient conditions.

The latest product of the Cytomat 10 series is based on the Cytomat 10 C automated incubator. It can be seamless integrated with existing automated assay hardware and provides the same fast access time of less than 10 seconds.

The Cytomat 10 Hotel (PH) offers multiple transfer positions, dual access point (DAP) and as combined benchtop-under bench solution, it allows flexible placement within the laboratory.

It is available with metal cover (STD; standard) and a perspex housing (PH) as a dust and light cover (option).

Matches your Capacity with 10 Stacker
  • 280pcs. 1536-well plates
  • 210pcs. 96-well or 384-well plates
  • 150pcs. transwell or insert plates
  • 100pcs. deep-well

Further stacker types on request
Mix and match different pitch stackers


Fast
  • Below 10sec. access time

Flexible Stacker Design
  • Compatible with existing Cytomat stacker
  • Standard or customized stacker pitch sizes
  • Mix and match different pitch stackers
  • Autoclaveable, stainless steel
  • Ventilation slots allow accurate temperature distribution

Convenient – Easy to use
  • Internal and direct barcode reader
  • Various transfer positions (up to 12) and dual access points (DAP)

Easy to integrate
  • Intuitive software commands
  • SILA (Standard in Lab Automation) Compliant
  • Multiple transfer stations

Certification
  • CE
  • RoHS
  • Manufactured in ISO 9001 and ISO 13485 accredited facility
Specifications
Access TimeBelow 10 sec.
Certifications/ComplianceCE Marked
Depth (English)33.0 in.
Depth (Metric)840 mm
Electrical Requirements100/230 V, 50/60 Hz
FeaturesFast Ambient and Random Access Storage, Perspex Housing as Light and Dust Cover, Dual Access Point (DAP)
Height (English)31.6 in.
Height (Metric)803 mm
Holds280 Pc. 1536-Well Plates, 210 Pc. 96-Well or 384-Well Plates, 150 Pc. Transwell or Insert Plates, 100 Pc. Deep-Well
ModelPerspex Housing
TypeAmbient Storage
Width (English)27.6 in.
Width (Metric)701 mm
Chamber MaterialStainless Steel
MaterialStainless Steel
Temperature Range (Metric)Ambient
Voltage100/230 V
Unit SizeEach