Accessories for Thermo Scientific™ Large Capacity Centrifuges
Actual product may vary
Thermo Scientific™
Accessories for Thermo Scientific™ Large Capacity Centrifuges
Complete your system with accessories for your Thermo Scientific™ large capacity centrifuge.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
50148262Scanner Holder
76007514Barcode Laser Scanner
75007730Drain Box (600 x 400 x 50mm)
Catalog number 50148262
Price (INR)
-
Description:
Scanner Holder
Thermo Scientific™ large capacity centrifuges are designed for high-throughput processing with the flexibility to select higher capacity rotors or match existing workflows.
Accessories designed to complete your Thermo Scientific large capacity centrifuge system.
Specifications
DescriptionScanner Holder
For Use With (Equipment)Thermo Scientific Cryofuge 8 and 16, Sorvall BP 8 and 16, and Sorvall BIOS 16 centrifuges
Product TypeScanner Holder
Unit SizeEach