Color-coded Sample Alert Labels
Color-coded Sample Alert Labels
Color-coded Sample Alert Labels
Color-coded Sample Alert Labels
Color-coded Sample Alert Labels
Thermo Scientific™
Color-coded Sample Alert Labels
Increase the safety and efficiency of sample handling and identification with color-coded sample alert labels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Type
503-0007BlueSample Label, Hydrochloric Acid
503-0004YellowSample Label, Sulfuric Acid
Catalog number 503-0007
Price (INR)
-
Request A Quote
Color:
Blue
Product Type:
Sample Label, Hydrochloric Acid
Increase the safety and efficiency of sample handling and identification with Thermo Scientific™ Color-Coded Sample Alert Labels. Each label is color-coded with the chemical preservative type for quick reference and safe handling. Sticks securely to plastic or glass sample containers.
  • Color-coded adhesive “dot“ with chemical preservative or short holding time alert
  • Roll of 1000 labels
  • Provides easy sample aliquot identification and enhances safe handling information visibility
  • Sticks securely to plastic or glass
Specifications
Certification ParametersNone
Height (English)1 in. (round)
Height (Metric)25.4 mm
No. per Case1,000
Certifications/ComplianceNone
ColorBlue
Product TypeSample Label, Hydrochloric Acid
Unit SizeEach