SR-3000 Solvent Rack without Degassers for UltiMate™ 3000 Pumps
SR-3000 Solvent Rack without Degassers for UltiMate™ 3000 Pumps
SR-3000 Solvent Rack without Degassers for UltiMate™ 3000 Pumps
Thermo Scientific™
SR-3000 Solvent Rack without Degassers for UltiMate™ 3000 Pumps
Secure large volumes of multiple eluents on this high capacity solvent rack. It supports low pressure gradient (LPG) systems with internal degassers.
Have Questions?
Catalog NumberVoltage
5035.920030 V
Catalog number 5035.9200
Price (INR)
-
Voltage:
30 V
Securely organize up to nine 1L eluent bottles using the Thermo Scientific™ SR-3000 Solvent Rack without desgassers. Placed on top of your Thermo Scientific™ Dionex™ UltiMate™ 3000 system stack, the solvent rack without a degasser supports pumps with integrated degassers. It is recommended in combination with all UltiMate 3000 Quaternary RSLC, Standard, and Micro Pumps with internal degassers (LPG-3400RS/SD/AB/M/MB). Its compact design conserves valuable bench space. The tray is resistant to the most commonly used HPLC solvents and buffer solutions and enables easy cleaning and maintenance.
Specifications
For Use With (Equipment)UltiMate 3000 Pumps
Voltage30 V
Weight (Metric)3 kg
TypeSolvent Rack
Unit SizeEach