Product Image
Applied Biosystems™

BRKT, System Power Supply

Have Questions?
Catalog NumberQuantity
50541 unit
Catalog number 5054
Price (INR)
-
Quantity:
1 unit
This is a spare part for the 310 Genetic Analyzer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)310 Genetic Analyzer
Product TypeSystem Power Supply
Quantity1 unit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.