UltiMate™ 3000 Fluorescence Detectors
Thermo Scientific™

UltiMate™ 3000 Fluorescence Detectors

Develop highly selective methods with sensitivity three to six orders of magnitude greater than UV detection with this fluorescence detector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDetection ModeModel
5078.0015Single ChannelFLD-3100 w/ Dual PMT
5078.00204 ChannelsFLD-3400 RS
5078.0010Single ChannelFLD-3100
Catalog number 5078.0015
Price (INR)
-
Detection Mode:
Single Channel
Model:
FLD-3100 w/ Dual PMT

Develop highly selective methods for trace analytes or novel molecules with sensitivity three to six orders of magnitude greater than UV detection. The Thermo Scientific™ Dionex™ UltiMate™ 3000 Fluorescence Detectors offer four channel acquisition at up to 200Hz so you can optimize quantification on the most sensitive channel and single-channel detection at up to 100Hz. 2D or 3D synchro, excitation, emission scans, zero order mode, or multichannel acquisition offer flexibility for method development or routine sample characterization. A variable emission filter allows real-time, compound-related sensitivity optimization.

Recommended Use

The Thermo Scientific™ UltiMate 3000 Rapid Separation Fluorescence Detector FLD-3400 is recommended for method development labs that need flexibility in detecting novel compounds that don’t absorb in the UV-vis region of the spectrum. It is ideal for the analysis of carbohydrates, glycans, and polyaromatic hydrocarbons (PAHs) in combination with any Thermo Scientific™ Dionex™ UltiMate 3000 system. The ability to cleave and derivitize glycans from intact proteins or monoclonal antibodies eliminates interferences from coeluting matrix peptides.

Sensitive Fluorescence Performance

  • Data rates at up to 200Hz detect the narrowest peaks in UHPLC separations.
  • Maximum stray light suppression provides excellent sensitivity.
  • Low limits of detection are possible with a Raman S/N: >550 ASTM (>2100 using dark signal as noise reference).
  • Active flow cell temperature control ensures stable fluorophore activity independent of ambient temperature.
  • Long-life xenon flash lamp delivers the highest sensitivity and long-term operation.
  • Optional second photomultiplier (PMT) for Dual-PMT operation extends the wavelength range up to 900nm without sacrificing sensitivity in the standard wavelength range.

Easy-to-Use Fluorescence Detector

  • Large front panel display allows easy monitoring of detector status.
  • Flow cell syringe injection kit enables cost-effective off-line measurements.
  • Flow cell optimized for trace analysis with narrow-bore UHPLC and conventional HPLC columns.
  • Automate qualification monitoring and tracking of flow cells for regulatory compliance.
  • Thermo Scientific™ Dionex™ Chromeleon™ Chromatography Data System software offers performance optimization and instrument wellness monitoring for ease of use and maximum uptime.
Specifications
Descriptionwith Dual-PMT
Detection ModeSingle Channel
DisplayLCD Monitor
FilterFixed
Flow CellWithout Flow Cell
Flow Cell Max. Pressure290 psi
Flow Cell Thermostatting15°C Above Ambient -50°C
Flow Cell Volume8 μL
GLP FeaturesAutomatic Instrument Qualification (AutoQ), System Wellness and Qualification Monitoring with Chromeleon Software, Lamp and Cell ID Chips.
LampXenon Flash Lamp
Lamp Pulse Frequency300 Hz
ModelFLD-3100 w/ Dual PMT
No. of Channels1
PC ConnectionUSB
Sensitivity>550 ASTM
Spectral Bandwidth20 nm
Temperature ControlYes
Wavelength Accuracy±2 nm
Wavelength Range200 to 880 nm
Weight (Metric)18 kg
Data Collection Rate100 Hz Max.
Detector TypeHigh-Sensitivity Fluorescence
TypeFluorescence Detector
Unit SizeEach