Cytomat™ 2 C470-LiN Series Automated Incubators
Thermo Scientific™

Cytomat™ 2 C470-LiN Series Automated Incubators

Temperature Range: Ambient +4°C to 70°C; Relative Humidity: <95%

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
51033039120 V
51033038100 V
51033040230 V
Catalog number 51033039
Price (INR)
-
Voltage:
120 V

Ensure a safe, reproducible process, unsurpassed temperature uniformity and stability with the next generation of mid-range Thermo Scientific™ Cytomat™ 2 C470-LiN series automated incubators for incubation/storage of cell culture applications.

These Cytomat incubators are available in various temperature ranges and gate positions for the hand-off to the lab automation robot. The device can be seamlessly integrated with a mean access time of 12 sec.

With a fully automated thermal decontamination and optional controlled humidity, the Cytomat 2 470-LiN series provides a new standard in significantly reducing the risk of contamination.

The Cytomat 2 C470-LiN series of automated incubators provide an ideal safe environment for storage and incubation of assays. This series of mid-range incubators offer a choice of temperature ranges (4 to 70°C), high humidity up to 95% r.H. stable CO2 conditions, and automated ContraCon decontamination routine without harmful gases.

The external cooling and heating unit is designed to allow the user to create a customized working space for easy system integration into any automated assay system.

Available options

  • Integrated barcode reader (1D) enables sample tracking, barcode transmission, and inventory management.
  • P3-Plate detection in stacker helps to eliminate labware handling errors.
  • HydraSmart Technology for precise humidity control without heat extraction and external water tank to avoid contamination.
  • Right speed setting for the right application with individual speed setting per plate and adjustable incremental speed settings (Slow/Medium/Fast).
  • Flexible positioning inside your lab with 6 possible gate positions including under bench in a single body.
  • Solid Copper inner chamber for natural contamination prevention.
  • Tower Shaker (ToS) for TRUE orbital shaking of applications requiring sample agitation and cells that need to be kept in suspension.
Specifications
Access Time12 sec.
Certifications/ComplianceCE Marked
Depth (English)20.1 in.
Depth (Metric)511mm
Electrical Requirements120V 60 Hz
FeaturesIncubation and storage unit with temperature range above; Humidity up to 95% r.H. CO2 range of 0-20 Vol. %; Includes decontamination routine-ContraCon
For Use With (Application)Incubation/storage for cell culture applications 4 to 70°C
Height (English)35.6 in.
Height (Metric)905mm
Holds56 Pc. 1536-Well Plates, 42 Pc. 96-Well or 384-Well Plates, 30 Pc. Transwell or Insert Plates, 20 Pc. Deep-Well
ModelCytomat 2 C470-LiN 120V 60Hz
TypeAutomated Incubator
Weight (Metric)90kg
Width (English)22.5 in.
Width (Metric)572mm
Chamber MaterialCopper
Relative Humidity<95%
Temperature Range (Metric)Ambient +4° to 70°C
Voltage120 V
Unit SizeEach