Product Image
Thermo Scientific™

Multiskan™ FC Microplate Photometer

Multiskan FC microplate photometer is a reliable instrument for a wide variety of research and routine applications. It reads 96- and 384-well plates and is equipped with shaking and incubation capabilities for temperature critical assays. It can be used as a stand-alone instrument or under PC control with intuitive Thermo Scientific SkanIt™ software.
Have Questions?
Catalog NumberMeasurement Speed
511191807 s for 96-well Plate, 13 s for 384-well Plate
Catalog number 51119180
Price (INR)
-
Measurement Speed:
7 s for 96-well Plate, 13 s for 384-well Plate
Multiskan FC brings together over 40 years experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard. A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.
  • Proven performance and reliable day-to-day results through patented optical design and in-built self-diagnostics
  • A broad wavelength range of 340–850 nm for a wide variety of research and routine applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves
  • Equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 nm and 620 nm, preinstalled
  • A comprehensive range of easy-to install additional filters available for order
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • Shaking and incubation up to 50°C for temperature critical assays
  • Ease of use through the large color screen and a variety of language versions
  • Visual and logical SkanIt software for comprehensive instrument control and data handling
  • Multiskan verification plate available for instrument performance verification
  • Robot compatibility for high-throughput environments
Specifications
Accuracy (Photometric)± 1% (0 to 3 Abs)or ±0.003 Abs, 450 nm
CompatibilityRobot
Depth (Metric)400 mm
DescriptionMultiskan FC with Incubator (China)
Dimensions (L x W x H)400 x 290 x 220 mm (15.7 x 11.4 x 8.7 in.)
For Use With (Application)ELISA, enzyme activity, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves
Height (Metric)220 mm
IncludesSkanIt software, Reserach Edition
Linearity0 to 3 Abs, ±2% at 450 nm
Measurement Speed7 s for 96-well Plate, 13 s for 384-well Plate
PrecisionCV ±0.2% (0.3 to 3 Abs), 450 nm
Software Language VersionsEnglish, German, French, Spanish, Portuguese, Russian, Chinese, Japanese
Warranty1 Year
Wavelength Range340 to 850 nm
Weight (Metric)8.5 kg
Width (Metric)290 mm
CuvettesNo
Incubator TemperatureAmbient +4°C to 50°C
InterfacePC Control, PC Software (Thermo Scientific SkanIt Software)
Light SourceQuartz Halogen Lamp
Measurement TechnologyAbsorbance
Plate Type96 and 384-well Plates
Product LineMultiskan
ShakingLinear
Software TypeInternal Software or PC Control with SkanIt Software
Wavelength SelectionFilters
Unit SizeEach

Frequently asked questions (FAQs)

Is Multiskan FC integrateable with robotics?

Yes, the Multiskan FC is intergrateable with robotics.

My Multiskan FC needs a new lamp. What is the catalog or part number? Can I replace it myself?

Yes, the Halogen lamp 6V/10W is Cat. No. 1410101. You can replace the lamp yourself using instructions in the Multiskan FC User Manual: https://www.thermofisher.com/order/catalog/product/51119100

I think my Multiskan FC isn't working right, but there's no error message. How can I check?

If the results are not as expected, performance can be verified using Multiskan Verification Plate (Cat. Nos. 24072800 (for 8 wavelengths, use with either model) or 24072805 (for 16 wavelengths, use with 51119100)).

Do I need to calibrate my Multiskan FC?

Readers are factory-calibrated and perform self-diagnostics on start-up to ensure consistent results over time. Should an error message appear indicating calibration is needed, contact your local Thermo Fisher Scientific microplate instrument service group at thermofisher.com/contactus.

Can I use uDrop Plate with Multiskan FC?

uDrop Plate is not compatible with Multiskan FC. For UV applications, refer to Multiskan Sky: https://www.thermofisher.com/order/catalog/product/51119500