Product Image
Thermo Scientific™

Multiskan™ FC Microplate Photometer

Multiskan FC microplate photometer is a reliable instrument for a wide variety of research and routine applications. It reads 96- and 384-well plates and is equipped with shaking and incubation capabilities for temperature critical assays. It can be used as a stand-alone instrument or under PC control with intuitive Thermo Scientific SkanIt™ software.
Have Questions?
Catalog number 51119180ET
Price (INR)
-
Multiskan FC brings together over 40 years experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard. A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.
  • Proven performance and reliable day-to-day results through patented optical design and in-built self-diagnostics
  • A broad wavelength range of 340–850 nm for a wide variety of research and routine applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves
  • Equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 nm and 620 nm, preinstalled
  • A comprehensive range of easy-to install additional filters available for order
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • Shaking and incubation up to 50°C for temperature critical assays
  • Ease of use through the large color screen and a variety of language versions
  • Visual and logical SkanIt software for comprehensive instrument control and data handling
  • Multiskan verification plate available for instrument performance verification
  • Robot compatibility for high-throughput environments
Specifications
Accuracy (Photometric)± 1% (0 to 3 Abs)or ±0.003 Abs, 450 nm
CompatibilityRobot
Depth (Metric)400 mm
DescriptionMultiskan ET (China)
Dimensions (L x W x H)400 x 290 x 220 mm (15.7 x 11.4 x 8.7 in.)
For Use With (Application)Endotoxin assays
Height (Metric)220 mm
IncludesSkanIt software, Reserach Edition
Linearity0 to 3 Abs, ±2% at 450 nm
Measurement Speed7 s for 96-well Plate, 13 s for 384-well Plate
PrecisionCV ±0.2% (0.3 to 3 Abs), 450 nm
Software Language VersionsEnglish, German, French, Spanish, Portuguese, Russian, Chinese, Japanese
Warranty1 Year
Wavelength Range340 to 850 nm
Width (Metric)290 mm
CuvettesNo
Incubator TemperatureAmbient +4°C to 50°C
InterfacePC Control, PC Software (Thermo Scientific SkanIt Software)
Light SourceQuartz Halogen Lamp
Measurement TechnologyAbsorbance
Plate Type96 and 384-well Plates
Product LineMultiskan
ShakingLinear
Software TypeInternal Software or PC Control with SkanIt Software
Wavelength SelectionFilters
Unit SizeEach