Vacutherm™ Oven Accessories and Options
Actual product may vary
Thermo Scientific™
Vacutherm™ Oven Accessories and Options
Choose from a range of accessories and options for Vacutherm™ ovens to meet application-specific requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
51900195Digicon Temperature ControllerVT 6060 P,VT 6130 P
51900194Digicon Temperature ControllerVT 6060 M,VT 6130 M
51900193Pressure ControllerVT 6060 M/P and VT6130 M/P
51900196Digicon Temperature ControllerVT 6060 P
51900197Digicon Temperature ControllerVT 6130 P
51900209Thermicon Temp Program ControllerVT 6060 M,VT6130 M
51900008Weekly program timer, mechanicalVT 6060 M/P and VT6130 M/P
51900161Weekly program timer, digitalVT 6060 M/P and VT6130 M/P
51900298Eurotherm Temp control, RS 422/485VT 6060 M and VT 6130 M
51900297Eurotherm Temp controller, RS 232VT 6060 M and VT 6130 M
51900080DoorVT 6130 P
51900079DoorVT 6060 P
51900078Vacuum ChamberVT 6130 P
51900075Vacuum ChamberVT 6060 M
51900077Vacuum ChamberVT 6060 P
51900069Digital Pressure DisplayVT 6060 M/P and VT6130 M/P
51900071Door GasketVT 6060 M
51900072Door GasketVT 6130 M
51900081Central Monitoring ConnectionVT 6060 M/P and VT6130 M/P
51900284RS 232 Computer InterfaceVT 6060 M/P and VT6130 M/P
51900326Precision Control ValveVT 6025
50043975ShelfVT 6060 M
50029890Support FrameVT6060 M/P
50046860Vacuum Connection KitVT 6025/6060/6130 M/P
50048621Stainless-steel trayVT6060 M/P
Catalog number 51900195
Price (INR)
-
Type:
Digicon Temperature Controller
For Use With (Equipment):
VT 6060 P,VT 6130 P
Choose from a range of Thermo Scientific™ Vacutherm™ Oven Accessories and Options to meet application-specific requirements.
Specifications
DescriptionDigicon™ S Temperature controller, 300°C
For Use With (Equipment)VT 6060 P,VT 6130 P
Temperature (Metric)300°C
TypeDigicon Temperature Controller
Unit SizeEach