Product Image
Thermo Scientific™
O2 Control (5 to 90%) with 3 door inner gas tight screen door
For Use With: Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
Have Questions?
Catalog NumberOxygen Control
519011465 to 90%
Catalog number 51901146
Price (INR)
-
Oxygen Control:
5 to 90%
Upgrade your incubators to take advantage of O2 control.
Specifications
Description5 to 90% O2 control with 3 door inner gas tight screen door
For Use With (Equipment)Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
LightingLED
Oxygen Control5 to 90%
Type5 to 90% O2 Control with 3 door inner gas tight screen door
Unit SizeEach