Electrochemical Detector Cells
Actual product may vary
Thermo Scientific™
Electrochemical Detector Cells
Choose from an extensive selection of electrochemical cells which have been optimized for a variety of critical applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
55-0685Coulometric, Flow-Through
55-0417Preanalytical, Flow-Through
Catalog number 55-0685
Price (INR)
-
Type:
Coulometric, Flow-Through
Choose from this selection of electrochemical cells which have been optimized for a variety of critical applications. Thermo Scientific™ Dionex™ Electrochemical Detector Cells for LC are unique in their configuration and performance. Our advanced sensor designs assure long-lasting, trouble-free performance.
For Use with Thermo Scientific Dionex Electrochemical Detectors
These Electrochemical Cells are recommended for use with the following detectors, which are compatible with all Thermo Scientific Dionex UHPLC+ systems:
  • Dionex™ Coulochem™ III Electrochemical Detector
  • Dionex™ CoulArray™ Coulometric Array Detector
Specifications
DescriptionCoulometric Array Cell, Model 6210
Detection ModeDC
For Use With (Equipment)Dionex CoulArray Coulometric Array Detector
No. of Channels4
Reference ElectrodePd
Working ElectrodesPG
Detector TypeElectrochemical (ECD)
TypeCoulometric, Flow-Through
Unit SizeEach