Axiom™ Apple Genotyping Array
Applied Biosystems™

Axiom™ Apple Genotyping Array

Axiom Apple Genotyping Array (Axiom_Apple480) was designed through the Affymetrix Expert Design Program in collaboration with the FruitBreedomics consortium (www.FruitBreedomics.com).Read more
Have Questions?
Catalog NumberQuantity
5505731 plate
Catalog number 550573
Price (INR)
1,667,818.00
1 plate
Quantity:
1 plate
Price (INR)
1,667,818.00
1 plate
Axiom Apple Genotyping Array (Axiom_Apple480) was designed through the Affymetrix Expert Design Program in collaboration with the FruitBreedomics consortium (www.FruitBreedomics.com). The sequencing and marker selection was conducted by experts from Fondazione Edmund Mach, INRA, Dalhousie University, Wageningen UR, University Di Bologna, and Universita Degli Studi Di Milano.

The 96-format array includes markers identified using whole-genome sequence data from 63 Malus domestica cultivars and two double haploid accessions. Table 1 lists the number of cultivars and corresponding country of origin used in the SNP discovery process.

Axiom Apple Genotyping Array includes 480,000 markers and together with Axiom Analysis Suite software overcomes the genotyping challenges associated with polyploidy, rapid linkage disequilibrium (LD) decay, and high polymorphism observed in the apple genome.

Array highlights
• Very high diversity
• Includes markers discovered in 63 worldwide M. domestica cultivars• High resolution to address rapid decay of LD   - 487,249 markers on the array
   - Bias towards common variants with minor allele frequency (MAF) >0.05
   - Includes 21,463 previously validated markers: 19,990 markers from an existing in-;market 20K Fruitbreedomics apple array and 1,473 markers identified using genotyping-by-sequencing (GBS)
• Absence of paralogous variants through the use of double haploid accessions in SNP discovery

Applications of Axiom Apple Genotyping Array
• Construction of high-resolution genetic maps
• Fine mapping of quantitative trait loci
• Genome-wide association studies

Required Products
GeneChip Command Console Software
GeneTitan Multi-Channel InstrumentAxiom Apple Genotyping Array (Axiom_Apple480) was designed through the Expert Design Program at Affymetrix in collaboration with the FruitBreedomics consortium (www.FruitBreedomics.com). The sequencing and marker selection was conducted by experts from Fondazione Edmund Mach, INRA, Dalhousie University, Wageningen UR, University Di Bologna, and Universita- Degli Studi Di Milano.

The 96-format array includes markers identified using whole-genome sequence data from 63 Malus domestica cultivars and two double haploid accessions. Table 1 lists the number of cultivars and corresponding country of origin used in the SNP discovery process.

Axiom Apple Genotyping Array includes 480,000 markers and together with Axiom™ Analysis Suite software overcomes the genotyping challenges associated with polyploidy, rapid linkage disequilibrium (LD) decay, and high polymorphism observed in the apple genome.

Array highlights
  • Very high diversity
  • Includes markers discovered in 63 worldwide M. domestica cultivars
  • High resolution to address rapid decay of LD
    • 487,249 markers on the array
    • Bias towards common variants with minor allele frequency (MAF) >0.05
    • Includes 21,463 previously validated markers: 19,990 markers from an existing in-market 20K Fruitbreedomics apple array and 1,473 markers identified using genotyping-by-sequencing (GBS)
  • Absence of paralogous variants through the use of double haploid accessions in SNP discovery
Applications of Axiom Apple Genotyping Array
  • Construction of high-resolution genetic maps
  • Fine mapping of quantitative trait loci
  • Genome-wide association studies

Table 1: The country of origin and the number of cultivars used in the SNP discovery process for Axiom Apple Genotyping Array.
Country of origin Number of cultivars
Australia 1
Central Europe 39
Canada 1
Iran 1
Simmental 16
Japan 1
New Zealand 1
Northern Europe 4
Russia 9
Tunisia 1
United States 5
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesApple
Unit Size1 plate

Frequently asked questions (FAQs)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.