Axiom™ Porcine Breeders Array, 384HT format
Applied Biosystems™

Axiom™ Porcine Breeders Array, 384HT format

The Applied Biosystems Axiom Porcine Breeders Array includes markers focused on genomic selection of commercial porcine breeds, including Large White,Read more
Have Questions?
Catalog NumberQuantity
5508331 plate
Catalog number 550833
Price (INR)
-
Quantity:
1 plate
The Applied Biosystems Axiom Porcine Breeders Array includes markers focused on genomic selection of commercial porcine breeds, including Large White, Landrace, Duroc, and Pietrain. Core markers for the array were selected from markers on the Axiom Porcine High-Density Array that were evaluated using samples from multiple commercial and domestic breeds, including European wild boars. The inclusion of the most informative and well-performing markers from existing porcine arrays ensures that this array can be used for comparison with data generated by previous studies. With the availability of the Axiom Long Format Export Tool, which converts Axiom array data into long format, the array also provides the ability to continue existing projects with no change to the current bioinformatics infrastructure and pipeline.

Features include:
• 55,150 markers for genomic and trait selection
• 42 trait-specific markers selected by United States Department of Agriculture
• 64 parentage markers, including ISAG core markers
• Evenly distributed SNPs across the porcine genome
• Marker annotations aligned to Sscrofa10.2/susScr3 genome and dbSNP 148
• Targeted for commercial breeds including Large White, Landrace, Duroc, and Pietrain

Applications
• Parentage analysis
• Identification of economically important traits
• Genetic improvement of pure lines

Required products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
Axiom Analysis Suite Software
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Product LineAxiom
Quantity1 plate
Shipping ConditionRoom temperature
ArrayDNA
Format384 HT-array Plate
Number of Arrays384 arrays
SpeciesPorcine
Unit Size1 plate
Contents & Storage
One plate containing 384 arrays, Store in refrigerator (2-8°C)

Frequently asked questions (FAQs)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.