Axiom™ BreedWheat 35K Genotyping Array (384HT format)
Applied Biosystems™
Axiom™ BreedWheat 35K Genotyping Array (384HT format)
The Axiom BreedWheat 35K Genotyping Array (Axiom TaBW35K) was designed in the framework of the BreedWheat Project in collaboration withRead more
Have Questions?
Catalog NumberQuantity
5508681 plate
Catalog number 550868
Price (INR)
1,357,029.00
1 plate
Quantity:
1 plate
Price (INR)
1,357,029.00
1 plate
The Axiom BreedWheat 35K Genotyping Array (Axiom TaBW35K) was designed in the framework of the BreedWheat Project in collaboration with Dr. Etienne Paux, project leader at the National Institute for Agricultural Research, Clermont-Ferrand, France. The BreedWheat Project, a French public-private partnership, aims at strengthening the competitiveness of the French wheat-breeding sector, as well as to address the societal demands for sustainability, quality, and safety in agricultural production. This array is also available in mini-96 format.
The 34,746 SNPs on this array were derived from the Axiom TaBW280K Genotyping Array (1). Several selection criteria were applied, including clustering profiles, even distribution (based on linkage disequilibrium data rather than physical positions), and known position on the IWGSC RefSeq v1.0.
Information on the SNPs, including physical and genetic positions, context sequences, and statistics derived from a panel of 96 wheat lines representative of the worldwide diversity are also provided.
Required products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
Axiom Analysis Suite Software
Reference
1. Rimbert H, Darrier B, Navarro J, Kitt J, Choulet F, et al. (2018) High throughput SNP discovery and genotyping in hexaploid wheat. PLOS ONE 13(1): e0186329.
2. Lange TM, Heinrich F, Enders M, Wolf M, Schmitt AO. (2020) In silico quality assessment of SNPs—A case study on the Axiom Wheat genotyping arrays. Current Plant Biology 21:100140.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 plate
Shipping ConditionRoom temperature
ArrayGenotyping
Format384 HT-array Plate
SpeciesWheat
Unit Size1 plate
Contents & Storage
One plate containing 384 arrays, Store in refrigerator (2-8°C)