Pharma 24 Twin-Screw Extruder
Pharma 24 Twin-Screw Extruder
Actual product may vary
Pharma 24 Twin-Screw Extruder
Pharma 24 Twin-Screw Extruder
Pharma 24 Twin-Screw Extruder
Pharma 24 Twin-Screw Extruder
Pharma 24 Twin-Screw Extruder
Pharma 24 Twin-Screw Extruder
Speed up and enhance continuous processing in pharmaceutical research, development or production with the Pharma 24 Twin-Screw Extruder.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
553-2038HME Application Set
553-2037TSG Application Set
553-2039Pharma 24 Extruder
Catalog number 553-2038
Price (INR)
-
Request A Quote
Description:
HME Application Set
The Thermo Scientific™ Pharma 24 Twin-Screw Extruder, with available HME and Twin-Screw Granulation (TSG) Application Sets, is ideal for continuous processing in research, development or production in the pharmaceutical industry. The Pharma 24 extruder drives reduced clean room area, consistent and controllable processing, reduced product changeover time, on-demand production, minimized scale-up risk and reliable cleaning. It is suitable for process analytical technology (PAT).
The Pharma 24 Twin-Screw Extruder features:
  • A small footprint to take up less space in clean rooms
  • Continuous operation which consumes fewer utilities than traditional batch processes
  • High free volume screw design
  • A small process inventory
  • Lower ingredient consumption which reduces handling and operator exposure
  • Segmented screws and barrel
  • Removable screws and liners
PAT Compatibility: PAT can be applied to your process to monitor parts of theprocess such as homogeneity of mixing or moisture content to assure required product quality. The Pharma 24 Extruder can be used with a range of PAT instruments such as Thermo Scientific™ FT-NIR system.
Options: We offer a full range of GMP compliant ancillaries which are matched to the extruder performance. This downstream equipment can be integrated into the extruder control system.
For TSG configurations: available ancillaries include volumetric or gravimetric dosing feeders and liquid injection pumps.
In addition, self-contained units of either configuration can be operated with recirculating chillers to reduce water consumption.
TSG configuration recommended for:
  • Wet granulation
  • Dry/melt granulation
For HME configurations: available ancillaries include volumetric or gravimetric dosing feeders, air cooled conveyors for product cooling, strand and face-cut pelletizers to make pellets, or chill roll/flaker units for rapid product cooling and flaking.
HME configuration recommended for:
  • Solubility enhancement
  • Modified release
  • Co-extrusion
  • Drug delivery systems
  • Biomedical products
Specifications
Barrel Bore Diameter24 mm
DescriptionHME Application Set
Dimensions (L x W x H)2075 x 875 x 1630 mm
Extruder DesignBarrel designed with removable top and bottom liners made from pharma grade steel, horizontally split, with top half lifting upward. Main feed port (cooled) plus 7 multi-purpose top feed/vent/injection ports
InterfaceTouch screen control panel, Siemens PLC, Standard HMI languages: German and English (optional languages on request), 21 CFR part 11 compliance, Four different user levels with predefined access rights
Max. Torque52.5 Nm/Screw
Power Connections400 V 3PH+N+PE, 50/60 Hz
Power Consumption50 A
Unit SizeEach