Pharma <i>mini</i> HME Micro Compounder Accessories
Actual product may vary
Thermo Scientific™
Pharma mini HME Micro Compounder Accessories
Accessories for the Pharma miniHME Micro Compounder used for Hot Melt Extrusion in research and development in the pharmaceutical industry
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
553-6120Batch Kit
553-6100Set of Co-rotating Screws
553-6105Set of Counter-rotating Screws
553-6400Manual Feeder
553-6410Mini Force Feeder
553-8001Documentation Package
553-0620CaliCut Post-extrusion System
Catalog number 553-6120
Price (INR)
-
Description:
Batch Kit
Accessories for the Pharma miniHME Micro Compounder used for Hot Melt Extrusion in research and development in the pharmaceutical industry.
Thermo Scientific™ CaliCut™ Post-extrusion System highlights:
  • Control precision: a 1D laser diameter gauge adjusts and pulls the strand enabling the diameter to be adjusted within set tolerances. A nominal strand diameter of 0.2 mm – 6 mm can be produced in conjunction with a nominal length of 1 mm – 200 mm.
  • Maximize flexibility: available with 350 mm, 700 mm or even 1200 mm belt lengths for cooling to meet a broad range of polymer formulation and throughput needs. Two different cutting mechanisms, a rotary and a linear cutting blade, optimize final implant sizing for a wide variety of implant materials. Make up to 600 cuts/minute.
  • Be 21 CFR part 11 compliant: stainless steel housing conforms to GMP guidelines while the software is used to control user access levels secures data export, and provide audit support. Documentation for IQ/OQ is provided.
Specifications
DescriptionBatch Kit
Speed Range10 to 360 rpm
Unit SizeEach