D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length
Actual product may vary
Thermo Scientific™
D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'S™ Extended Length tips — superior quality with the flexibility to choose tips for any application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFilteredTip Style
5546TS
also known as 5546
300 μLNon-sterileNon-filteredLow Retention, Wide Bore, Extended Length
541830 μLNon-sterileFilteredLow Retention, Extended Length
5538300 μLSterileFilteredLow Retention, Extended Length
5548300 μLSterileFilteredLow Retention, Wide Bore, Extended Length
5561220 μLNon-sterileNon-filteredExtended Length
541630 μLNon-sterileNon-filteredLow Retention, Extended Length
550630 μLNon-sterileNon-filteredLow Retention, Extended Length
5536300 μLNon-sterileNon-filteredLow Retention, Extended Length
541730 μLSterileNon-filteredLow Retention, Extended Length
550730 μLSterileNon-filteredLow Retention, Extended Length
5537300 μLSterileNon-filteredLow Retention, Extended Length
5547300 μLSterileNon-filteredLow Retention, Wide Bore, Extended Length
Catalog number 5546TS
also known as 5546
Price (INR)
-
Volume (Metric):
300 μL
Sterility:
Non-sterile
Filtered:
Non-filtered
Tip Style:
Low Retention, Wide Bore, Extended Length
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Extended Length Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision.
  • Sterile, sterile filtered and wide-bore tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x2, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
Specifications
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
DisposableSingle-use
Packaging20 x 96 Tips
Quantity1920 tips
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus, Thermo Scientific Versette
Product LineD.A.R.T.S
SterilityNon-sterile
Tip StyleLow Retention, Wide Bore, Extended Length
Volume (Metric)300 μL
Unit SizeCase of 1920