Hamilton™ Reagent Reservoirs, 200 mL
Thermo Scientific™
Hamilton™ Reagent Reservoirs, 200 mL
These reagent reservoirs are for use with the NIMBUS Presto System. The special bottom geometry minimizes residual volumes. They areRead more
Have Questions?
Catalog NumberQuantity
56695-011 x 10 reservoirs
Catalog number 56695-01
Price (INR)
11,112.00
Each
Quantity:
1 x 10 reservoirs
Price (INR)
11,112.00
Each
These reagent reservoirs are for use with the NIMBUS Presto System. The special bottom geometry minimizes residual volumes. They are made of polypropylene and manufactured under clean room conditions. They are certified CE and IVD-compliant.
Specifications
PackagingIndividually wrapped
Quantity1 x 10 reservoirs
Capacity (Metric)200 mL
MaterialPolypropylene
SterilityNon-sterile
TypeTrough
Unit SizeEach