HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System
HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System
HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System
HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System
HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System
Thermo Scientific™
HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System
HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System torque rheometer platform are focused on the analysis of rheological and processing characteristics of various materials.
Have Questions?
Catalog NumberTorque Rheometer
567-2110Yes
Catalog number 567-2110
Price (INR)
-
Request A Quote
Torque Rheometer:
Yes
HAAKE™ Rheomex QC lab Single Screw Extruders for the HAAKE™ PolyLab™ QC System torque rheometer platform are focused on the analysis of rheological and processing characteristics of various materials.
With only a small sample amount, customers can simulate production processes and generate films or profiles for further testing.
Single-screw extrusion
  • Proven single-screw laboratory extruders deliver reliable data captured during the extrusion process to verify process parameters (speed, energy, temperature) for unknown materials or to manufacture smaller quantities of a new polymer (as strands, sheets, pellets, blown films) during research and development. The extruders are equipped with measuring ports for melt pressure and melt temperature to study the process parameters along the extruder barrels. These barrels are available in different lengths (length/diameter ratio). A die can be connected to the end of the extruder barrel to form the polymer melt as strand or film. Special rheological dies (slit, rod, x-die) allow the determination of shear- and elongational viscosity at defined shear rates. A variety of screw geometries to customize the extrusion process with different compression ratios, venting and mixing sections is offered. Wear-reduced screws and barrels increase the lifetime of the system. Standard feeders for pellets and special feeding systems for powders, pastes, liquids or highly viscous rubber complete the set up.
    Typical single-screw investigations
    • Extrudability of newly developed materials
    • Manufacturing homogeneous melts
    • Manufacturing films, foils, strands and profiles for optical and mechanical testing as well as outdoor exposure tests
    • Measuring rheological behavior (viscosity, elasticity)
    • Testing individual and combined influences of additives (stabilizer, lubricant) and functional additives (anti-oxidation, UV-stabilizers, pigments and fillers)
    • Foam extrusion
    The HAAKE Rheomex measuring extruder contains the necessary measuring, control and monitoring technology for the respective application. A new concept simplifies handling in the laboratory and enables secure operation. This is characterized by:
    • Mobile measuring sensors avoid the necessity of handling of heavy and hot parts
    • Integrated heating and cooling circuits reduce the number of plug connections and thus the danger of making incorrect connections
    • Several peripheral units such as feeding and post-extrusion units are computer controllable thus meaning better reproducibility and documentation
  • Specifications
    DescriptionHAAKE PolyLab QC Single Screw Extruder
    Torque RheometerYes
    Unit SizeEach