CaptureSelect™ CH1-XL RoboColumn™ Pre-packed Column
Thermo Scientific™

CaptureSelect™ CH1-XL RoboColumn™ Pre-packed Column

Thermo Scientific™ CaptureSelect™ RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
5943462200200 μL
5943462600600 μL
Catalog number 5943462200
Price (INR)
-
Capacity (Metric):
200 μL

CaptureSelect RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, thereby enabling purification of all Fab fragments, independent of the light-chain type (kappa or lambda) and without binding of over-expressed light chain and light-chain dimers. In addition, it can be used for the purification of human IgGs from all four subclasses (1, 2, 3, and 4).

Features of this affinity matrix include:

  • Platform approach for purification of high quality and correctly assembled human Fab fragments
  • No co-purification of over-expressed free light chains due to selectivity for the CH1 domain
  • Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
  • Caustic cleaning with up to 50 mM NaOH
  • Excellent scalability
  • No cross-binding to bovine IgG, therefore presence of fetal bovine serum is not a problem
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL and 600 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 200 μL: 5 x 10 mm
  • Column dimension: 600 μL: 5 x 30 mm
  • Matrix: agarose based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect CH1-XL affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: >15 mg/mL human Fab fragment depending on flow rate, column height, contact time. For humanized Fabs, lower capacities may be observed.
  • Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0–4.5
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/h

RoboColumn is a trademark of Repligen.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity>15 mg/mL human Fab fragment
Column TypeRoboColumn
Diameter (Metric)5 mm
Flow Rate16 to 150 cm/hr.
For Use With (Application)Chromatography
Format8-well Strips
Length (Metric)10 mm
Ligand TypeCH1-XL affinity ligand
MatrixAgarose-based, epoxide-activated
Max. Operating Pressure2 bar
Quantity200 μL
Shipping ConditionRoom temperature, or on wet ice
Stationary PhaseCH1-XL affinity ligand on agarose bead
Capacity (Metric)200 μL
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeColumn
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C)