Air Filter Kits
Actual product may vary
Air Filter Kits
Air Filter Kits
Thermo Scientific™
Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeSize
598000000007ThermoFlex 2500Air and Fluid Filter35 micron
611000000089Merlin M25 or M33Air FilterN/A
611000000090Merlin M75Air FilterN/A
611000000091Merlin M100 or M150Air FilterN/A
598000000004ThermoFlex 900 or 1400Air and Fluid Filter100 micron
598000000005ThermoFlex 900 or 1400Air and Fluid Filter35 micron
598000000006ThermoFlex 2500Air and Fluid Filter100 micron
598000000008ThermoFlex 3500 or 5000Air and Fluid Filter100 micron
598000000009ThermoFlex 3500 or 5000Air and Fluid Filter35 micron
598000000014ThermoFlex 7500 or 1000Air and Fluid Filter100 micron
598000000013ThermoFlex 7500 or 1000Air and Fluid Filter35 micron
096270Polar Accel 250 and 500Air FilterN/A
Catalog number 598000000007
Price (INR)
-
For Use With (Equipment):
ThermoFlex 2500
Type:
Air and Fluid Filter
Size:
35 micron
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific Neslab HX, CFT or M series recirculating chillers.
Specifications
For Use With (Equipment)ThermoFlex 2500
Size35 micron
TypeAir and Fluid Filter
Unit SizeEach