Caps for Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers
Actual product may vary

Caps for Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers

Prevent bio-hazardous spills with these Caps for Thermo Scientific™ Samco™ Histology Bio-Tite™ Specimen Containers.
Have Questions?
Catalog NumberClosure SizeClosure Color
5A0032
also known as 5A 0032
35 mmWhite
Catalog number 5A0032
also known as 5A 0032
HSN 39233090
Price (INR)
156,551.00
CS
Closure Size:
35 mm
Closure Color:
White
Price (INR)
156,551.00
CS

Prevent bio-hazardous spills with these Caps for Thermo Scientific™ Samco™ Histology Bio-Tite™ Specimen Containers.

  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing

Ordering Information:

Bulk packed, one bag per case

Specifications
Product TypeSpecimen Container
Closure IncludedYes
ShapeCylindrical
SterilityNon-sterile
Capacity (Metric)20 mL
Closure ColorWhite
Closure Size35 mm
Closure TypeBio-Tite Lid
DescriptionCaps, 35mm; White
Diameter (Metric)35 mm
No. per Case4000/Cs.
No. per Pack4000/Pk.
Unit SizeCS