Specifications
Chemical Name or MaterialAcetone
DescriptionFerrule
For Use With (Equipment)LC Columns
MaterialStainless Steel
TypeHigh Pressure 1/16 in. Ferrule
CAS67-64-1
Quantity10/Pk.
ColorColorless
Health Hazard 2P210-P305 + P351 + P338-P370 + P378-P403 + P235
Density0.791g/mL at 25°C
RTECS NumberAL3150000
Quantity1 L
Explosion Limit0.132
Flash Point−17°C (1.4°F)
Molecular FormulaC3H6O
Linear FormulaCH3COCH3
MDL NumberMFCD00008765
Vapor Pressure184 mmHg (20°C)
UN NumberUN1090
Beilstein635680
Synonymacetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
IUPAC Namepropan-2-one
Molecular Weight (g/mol)58.08
PubChem CID180
ChEBICHEBI:15347
Formula Weight58.08g/mol
Percent Purity≥99.8% (GC)
GradeCHROMASOLV™
Unit SizeEach