HyperSep™ Pressure Manifold Air Filter
HyperSep™ Pressure Manifold Air Filter
Actual product may vary
HyperSep™ Pressure Manifold Air Filter
Prevent contaminants in the air source from entering the manifold system.
Have Questions?
Catalog NumberDescription
60104-273In Line Air Filter, 1 pk
Catalog number 60104-273
HSN 39269099
Price (INR)
189,855.00
Each
Add to cart
Description:
In Line Air Filter, 1 pk
Price (INR)
189,855.00
Each
Add to cart
Prevent contaminants from the air source from entering the system the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Installation Kit for all the parts needed to set up your manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
See related products if you need replacements for any of these HperSep Positive Pressure Manifold accessories.
Specifications
DescriptionIn Line Air Filter, 1 pk
For Use With (Application)SPE
For Use With (Equipment)Positive Pressure Manifold
Product LineHyperSep
Volume (Metric)In Line Air Filter
Unit SizeEach