SMART Digest™ Oligo Isolation Kit
Thermo Scientific™

SMART Digest™ Oligo Isolation Kit

Thermo Scientific™ SMART Digest™ Oligo Isolation Kits enable efficient digestion and extraction of oligonucleotide-based drugs and metabolites from biofluids for accurate quantitation by LC-MS. This kit includes all necessary enzymes, buffers, and solid phase extraction (SPE) materials.
Have Questions?
Catalog NumberVolume (Metric)
60180-1011 mL
Catalog number 60180-101
Price (INR)
-
Volume (Metric):
1 mL
The SMART Digest™ Oligo Isolation Kit provides a streamlined solution for the digestion and extraction of oligonucleotide-based drugs and metabolites from biofluids, ensuring precise quantitation by LC-MS. This kit includes all essential enzymes, buffers, and solid phase extraction (SPE) materials, offering a comprehensive and reliable choice for your laboratory needs.
  • Efficiently digests and extracts oligonucleotide-based drugs and metabolites from biofluids
  • Ensures precise quantitation by LC-MS
  • Includes all necessary enzymes for effective digestion
  • Comes with required buffers for optimal enzyme activity
  • Features solid phase extraction (SPE) components for high-throughput sample processing
  • Comprehensive kit designed to streamline your workflow
Specifications
EnzymeProteinase K
For Use With (Application)Digestion and extraction of oligonucleotide-based drugs and metabolites from biofluids prior to quantitation by LC-MS
IncludesSMART Digest Soluble Proteinase K, Proteinase K Digest Buffer, 10 mg, 1 mL, 96-well, WAX SPE Plate, 25 mL SPE Elution Buffer
Volume (Metric) Well1 mL
Bed Weight10 mg
CaptureNone
CleavageProteinase K cleaves aromatic, aliphatic, or hydrophobic residues in P1 and P1' positions at the carboxyl side of amino acid residues.
For Use With (Equipment)Universal
FormatNon-Magnetic
Particle Size30 to 40 μm
Product LineSMART Digest
SorbentMixed Mode Weak Anion Exchanger, Polymeric
TypeOligo Isolation
Volume (Metric)1 mL
Unit SizeEach