Search
Select from a range of quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters.
| Catalog Number | Type |
|---|---|
| 60180-817 | Triple Base |
| 60180-815 | Dual Base |
| 60180-837 | O-Rings for Baseplates |
| 60180-838 | Flush Cap |
| 60180-818 | Four-Position Base |
Use quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters. A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change. The baseplates have a one-time installation procedure. Super Clean Cartridge Filters are not included.