Click-On™ Inline Gas Filter Connectors
Actual product may vary
Click-On™ Inline Gas Filter Connectors
Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterialDiameter (English) Outer
60180-834Wall Mounting ClampNo Information AvailableN/A
60180-809End FittingBrass0.125 in.
60180-810Steel End FittingSteel0.125 in.
60180-811End FittingBrass0.25 in.
60180-812Steel End FittingSteel0.25 in.
60180-813Double Ended ConnectorNo Information AvailableN/A
60180-833Replacement Filter O-RingNo Information AvailableN/A
Catalog number 60180-834
Price (INR)
19,290.00
Pack
Type:
Wall Mounting Clamp
Material:
No Information Available
Diameter (English) Outer:
N/A
Price (INR)
19,290.00
Pack
Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters. The end fittings remain installed during filter replacement, thereby reducing the cost of clean gas.
  • Incorporates needle valves that close lines and prevent contamination
  • End fittings measure 0.125in. (0.3cm) or 0.25in. (0.6cm)
  • Reusable
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Diameter (English) OuterN/A
For Use With (Equipment)Click-On Inline Filter
MaterialNo Information Available
TypeWall Mounting Clamp
Unit SizePack